Benzhydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

A12268-14 A12268-22 A12268-36
CAAAAA12268-14EA 51.82 CAD
CAAAAA12268-14 CAAAA12268-22 CAAAA12268-36
Benzhydrazide 98%
Benzhydrazide

Purity: 98% CAS No.: 613-94-5 Molecular Formula: C7H8N2O Formula Weight: 136.15 Melting Point: 113-117° Beilstein Reference: 471797

Formula: C₇H₈N₂O
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00007596
CAS Number: 613-94-5
EINECS: 210-363-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR