1,8-Dihydroxy-9,10-anthraquinone 95%

Supplier: Thermo Scientific Chemicals
Warning

A12001-22 A12001-36
CAAAA12001-22EA 81.84 CAD
CAAAA12001-22 CAAAA12001-36
1,8-Dihydroxy-9,10-anthraquinone 95%
1,8-Dihydroxy-9,10-anthraquinone

MDL: MFCD00001211 Beilstein Registry No.: 2054727


Purity: 95% CAS No.: 117-10-2 Molecular Formula: C14H8O4 Formula Weight: 240.21 Melting Point: 191-193° subl. Beilstein Reference: 2054727 Merck Reference: 14,2815 MDL No.: MFCD00001211

Formula: C₁₄H₈O₄
Melting Pt: 191…193 °C (subl.)
Density: 1.537 g/cm³ (20 °C)
Flash Pt: ≥ 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00001211
CAS Number: 117-10-2
EINECS: 204-173-5
Merck Index: 14,2815

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR