(Diethylamino)sulfur trifluoride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DAST

A11992-06 A11992-14 A11992-22
CAAAA11992-06EA 172.22 CAD
CAAAA11992-06 CAAAA11992-14 CAAAA11992-22
(Diethylamino)sulfur trifluoride 95%
(Diethylamino)sulphur trifluoride

MDL: MFCD00000363 Beilstein Registry No.: 184906 Fieser: 6,183 12,183 13,110 16,128 18,135 19,126 20,142 21,170


Purity: 95% CAS No.: 38078-09-0 Molecular Formula: C4H10F3NS Formula Weight: 161.19 Storage Temperature: 2°C to 8°C Flash Point: 23°(73°F) Refractive Index: 1.4115 Boiling Point: 30-32°/3mm Beilstein Reference: 1849066 UN No.: 2920 MDL No.: MFCD00000363

Formula: C₄H₁₀F₃NS
MW: 161.19 g/mol
Boiling Pt: 30…32 °C (3 mmHg)
Density: 1.23
Flash Pt: 23 °C (73 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000363
CAS Number: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR