Trityl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A11799-14 A11799-22 A11799-36
CAAAA11799-14EA 49.61 CAD
CAAAA11799-14 CAAAA11799-22 CAAAA11799-36
Trityl chloride 98%
Trityl chloride

MDL: MFCD00000813 Beilstein Registry No.: 397363 Fieser: 1,1254 2,453 4,565 5,741 6,656 15,359 18,394 21,479


Purity: 98% CAS No.: 76-83-5 Molecular Formula: C19H15Cl Formula Weight: 278.78 Melting Point: 110-114° Boiling Point: 230-235°/20mm UN No.: 3261 MDL No.: MFCD00000813

Formula: (C₆H₅)₃CCl
Boiling Pt: 230…235 °C/20 mm
Melting Pt: 110…114 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR