4-Cholesten-3-one 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: (8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one, (+)-4-Cholesten-3-one

A11743-06 A11743-14
CAAAA11743-06EA 137.46 CAD
CAAAA11743-06 CAAAA11743-14
4-Cholesten-3-one 98+%
4-Cholesten-3-one

MDL: MFCD00003663 Beilstein Registry No.: 4707774 Optical Rotation: +93°C (c=1 in chloroform)


Purity: 98+% CAS No.: 601-57-0 Molecular Formula: C27H44O Formula Weight: 384.65 Melting Point: 79-81° Beilstein Reference: 4707774

Formula: C₂₇H₄₄O
MW: 384.65 g/mol
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003663
CAS Number: 601-57-0
EINECS: 210-005-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR