(2-Fluorophenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A11612-06 A11612-14 A11612-22
CAAAA11612-06EA 39.66 CAD
CAAAA11612-06 CAAAA11612-14 CAAAA11612-22
(2-Fluorophenyl)acetonitrile 97%
(2-Fluorophenyl)acetonitrile

MDL: MFCD00001897 Beilstein Registry No.: 1862361


Purity: 97% CAS No.: 326-62-5 Molecular Formula: C8H6FN Formula Weight: 135.14 Flash Point: 100°(212°F) Refractive Index: 1.5010 Boiling Point: 235° Beilstein Reference: 1862361 UN No.: 3276 MDL No.: MFCD00001897

Formula: C₈H₆FN
Boiling Pt: 235 °C
Density: 1.061
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001897
CAS Number: 326-62-5
EINECS: 206-313-0
UN: 2929
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR