Diisoamyl ether (mixture of isomers) 96%

Supplier: Thermo Scientific Chemicals
Warning

A11231-AC A11231-AE A11231-AP
CAAAA11231-ACEA 46.14 CAD
CAAAA11231-AC CAAAA11231-AE CAAAA11231-AP
Diisoamyl ether (mixture of isomers) 96%
Diisoamyl ether (mixture of isomers)

Purity: 96% CAS No.: 544-01-4 Molecular Formula: [(CH3)2CHCH2CH2]2O Formula Weight: 158.29 Flash Point: 46°(115°F) Refractive Index: 1.407 Boiling Point: 172-174° Beilstein Reference: 1698014 Merck Reference: 14,5118 UN No.: 3271 MDL No.: MFCD00008947

Formula: C₁₀H₂₂O
Boiling Pt: 172…174 °C
Density: 0.78
Flash Pt: 46 °C(115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008947
CAS Number: 544-01-4
EINECS: 208-857-4
UN: 1993
ADR: "3",III
Merck Index: 14,5118

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR