DL-Camphor 96%

Supplier: Thermo Scientific Chemicals
Warning

A10936-22 A10936-0C A10936-0E A10936-36
CAAAAA10936-22EA 53.09 CAD
CAAAAA10936-22 CAAAA10936-0C CAAAA10936-0E CAAAA10936-36
DL-Camphor 96%
DL-Camphor

MDL: MFCD00074738 Beilstein Registry No.: 1907611 Soluble in alcohols, ether, chloroform, benzene


Purity: 96% CAS No.: 76-22-2 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 64°(147°F) Physical Form: Crystalline powder or lumps Density: 0.992 Appearance: White Odor: Camphor-like Melting Point: 172-176° Boiling Point: 204° Solubility: Soluble in alcohols, ether, chloroform, benzene Beilstein Reference: 1907611 Merck Reference: 14,1732 UN No.: 2717 MDL No.: MFCD00074738

Formula: C₁₀H₁₆O
Boiling Pt: 204 °C
Melting Pt: 172…176 °C
Density: 0.992
Flash Pt: 64 °C (147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III
Merck Index: 14,1732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR