N-Boc-L-tyrosine 98+%

Supplier: Thermo Scientific Chemicals

A10810-03 A10810-06 A10810-14
CAAAA10810-03EA 33.34 CAD
CAAAA10810-03 CAAAA10810-06 CAAAA10810-14
N-Boc-L-tyrosine 98+%
N-Boc-L-tyrosine

Purity: 98+% CAS No.: 3978-80-1 Molecular Formula: C14H19NO5 Formula Weight: 281.31 Storage Temperature: 2°C to 8°C Melting Point: 141° dec. Beilstein Reference: 2816406 Optical Rotation: +37° (c=1 in dioxan) MDL No.: MFCD00037179

Formula: C₁₄H₁₉NO₅
Melting Pt: 141 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00037179
CAS Number: 3978-80-1
EINECS: 223-613-7

Order Now

Specification Test Results

Optical rotation +37° (c=1 in dioxan)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR