4,5-Dimethylthiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A10785-06 A10785-14 A10785-22
CAAAA10785-06EA 44.56 CAD
CAAAA10785-06 CAAAA10785-14 CAAAA10785-22
4,5-Dimethylthiazole 98%
4,5-Dimethylthiazole

MDL: MFCD00005336 Beilstein Registry No.: 105694


Purity: 98% CAS No.: 3581-91-7 Molecular Formula: C5H7NS Formula Weight: 113.18 Flash Point: 51°(123°F) Refractive Index: 1.5210 Boiling Point: 157-158° Beilstein Reference: 105694 UN No.: 1993 MDL No.: MFCD00005336

Formula: C₅H₇NS
Boiling Pt: 157…158 °C
Density: 1.070
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005336
CAS Number: 3581-91-7
EINECS: 222-703-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR