2-Chloro-α,α,α-trifluorotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

A10753-0E A10753-22 A10753-36
CAAAA10753-0EEA 352.34 CAD
CAAAA10753-0E CAAAA10753-22 CAAAA10753-36
2-Chloro-α,α,α-trifluorotoluene 99%
2-Chloro-α,α,α-trifluorotoluene

MDL: MFCD00000561 Beilstein Registry No.: 510993


Purity: 99% CAS No.: 88-16-4 Molecular Formula: C7H4ClF3 Formula Weight: 180.56 Flash Point: 58°(136°F) Refractive Index: 1.4560 Melting Point: -6° Boiling Point: 151-153° Beilstein Reference: 510993 UN No.: 2234 MDL No.: MFCD00000561

Formula: C₇H₄ClF₃
Boiling Pt: 151…153 °C
Melting Pt: –6 °C
Density: 1.369
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000561
CAS Number: 88-16-4
EINECS: 201-805-1
UN: 2234
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR