(3-Aminopropyl)triethoxysilane 98%

Supplier: Thermo Scientific Chemicals
Danger

A10668-0E A10668-22 A10668-36
CAAAA10668-0EEA 826.34 CAD
CAAAA10668-0E CAAAA10668-22 CAAAA10668-36
(3-Aminopropyl)triethoxysilane 98%
(3-Aminopropyl)triethoxysilane

MDL: MFCD00008207 Beilstein Registry No.: 1754988


Purity: 98% CAS No.: 919-30-2 Molecular Formula: C9H23NO3Si Formula Weight: 221.37 Flash Point: 104°(219°F) Refractive Index: 1.4210 Odor: Amine-like pH: 11 Boiling Point: 217° Beilstein Reference: 1754988 UN No.: 2735 MDL No.: MFCD00008207

Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
Boiling Pt: 217 °C
Melting Pt: < –70 °C
Density: 0.948
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR