Acetaldehyde oxime (mixture of syn and anti) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetaldoxime

A10640-18 A10640-30
CAAAA10640-18EA 214.88 CAD
CAAAA10640-18 CAAAA10640-30
Acetaldehyde oxime (mixture of syn and anti) 98%
Acetaldehyde oxime (mixture of syn and anti)

Fieser: 1,3


Purity: 98% CAS No.: 107-29-9 Molecular Formula: CH3CH=NOH Formula Weight: 59.07 Flash Point: 36°(96°F) Refractive Index: 1.4260 Odor: Characteristic Boiling Point: 115-117° Beilstein Reference: 1209252 Merck Reference: 14,42 UN No.: 2332 MDL No.: MFCD00002124

Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 115…117 °C
Density: 0.969
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002124
CAS Number: 107-29-9
EINECS: 203-479-6
UN: 2332
ADR: 3 III
Merck Index: 14,42

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR