5-Methylbenzotriazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10615-14 A10615-22 A10615-36
CAAAA10615-14EA 108.86 CAD
CAAAA10615-14 CAAAA10615-22 CAAAA10615-36
5-Methylbenzotriazole 98+%
5-Methylbenzotriazole

MDL: MFCD00005702 Beilstein Registry No.: 116658


Purity: 98+% CAS No.: 136-85-6 Molecular Formula: C7H7N3 Formula Weight: 133.15 Odor: Odorless pH: 5.3 Melting Point: 77-84° Boiling Point: 210-212°/12mm Beilstein Reference: 116658 MDL No.: MFCD00005702

Formula: C₇H₇N₃
Boiling Pt: 210…212 °C/12 mm
Melting Pt: 77…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005702
CAS Number: 136-85-6
EINECS: 205-265-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR