α,α,α-Trifluoro-3-tolualdehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23

A10463-06 A10463-14 A10463-22
CAAAA10463-06EA 84.85 CAD
CAAAA10463-06 CAAAA10463-14 CAAAA10463-22
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde

MDL: MFCD00003373 Beilstein Registry No.: 2327537


Purity: 97% CAS No.: 454-89-7 Molecular Formula: C8H5F3O Formula Weight: 174.12 Flash Point: 68°(154°F) Refractive Index: 1.4650 Odor: Almond-like Boiling Point: 83-86°/30mm Beilstein Reference: 2327537 MDL No.: MFCD00003373

Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 83…86 °C (30 mmHg)
Density: 1.304
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR