Molecular sieve 13X (1.0 nm, 10 Å), powder

Supplier: Thermo Scientific Chemicals

Synonyms: Linde 13X

A10378-0B A10378-0I A10378-30
CAAAA10378-0BEA 133.51 CAD
CAAAA10378-0B CAAAA10378-0I CAAAA10378-30
Molecular sieve 13X (1.0 nm, 10 Å), powder
Molecular sieve 13X (1.0 nm, 10 Å)

MDL: MFCD00131613


CAS No.: 63231-69-6 MDL No.: MFCD00131613

Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR