3',5'-Bis(trifluoromethyl)acetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A10341-06 A10341-14
CAAAA10341-03EA 64.18 CAD
CAAAA10341-03 CAAAA10341-06 CAAAA10341-14
3',5'-Bis(trifluoromethyl)acetophenone 98%
3',5'-Bis(trifluoromethyl)acetophenone

MDL: MFCD00009910 Beilstein Registry No.: 2384789


Purity: 98% CAS No.: 30071-93-3 Molecular Formula: C10H6F6O Formula Weight: 256.15 Flash Point: 82°(179°F) Refractive Index: 1.4230 Boiling Point: 85°/8mm Beilstein Reference: 2384789 MDL No.: MFCD00009910

Formula: C₁₀H₆F₆O
Boiling Pt: 85 °C/8 mm
Density: 1.422
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009910
CAS Number: 30071-93-3
EINECS: 250-023-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR