2-Bromo-α,α,α-trifluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-(trifluoromethyl)benzene, 2-Bromobenzotrifluoride

A10134-36 A10134-14 A10134-22
CAAAAA10134-36EA 440.82 CAD
CAAAAA10134-36 CAAAA10134-14 CAAAA10134-22
2-Bromo-α,α,α-trifluorotoluene 98%
2-Bromo-α,α,α-trifluorotoluene

Purity: 99% CAS No.: 392-83-6 Molecular Formula: C7H4BrF3 Formula Weight: 225.01 Flash Point: 68°(154°F) Refractive Index: 1.4820 Odor: Aromatic Boiling Point: 167-168° Beilstein Reference: 1945750 MDL No.: MFCD00000373

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 167…168 °C
Density: 1.662
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000373
CAS Number: 392-83-6
EINECS: 206-879-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR