Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75% in isopropanol (IPA)

Supplier: Thermo Scientific Chemicals
Danger

89360-22 89360-36
CAAA89360-22EA 55.3 CAD
CAAA89360-22 CAAA89360-36
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75% in isopropanol (IPA)
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV)

Ti 9.9%


CAS No.: 17927-72-9 Molecular Formula: C16H28O6Ti Formula Weight: 364.30 Flash Point: 18°(61°F) Physical Form: Liquid Density: 1.010 Refractive Index: 1.494 Sensitivity: Moisture Sensitive Appearance: Brown Odor: Alcohol-like UN No.: 1219 MDL No.: MFCD00000031

Formula: C₁₆H₂₈O₆Ti
Density: 1.010
Flash Pt: 18 °C (61 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000031
CAS Number: 17927-72-9
EINECS: 241-866-1
UN: 1219
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR