Barium titanate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

88266-14 88266-22
CAAA88266-14EA 162.74 CAD
CAAA88266-14 CAAA88266-22
Barium titanate ≥99.9% (metals basis)
Barium titanate

For vacuum deposition


Purity: 99.9% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 3-12mm Pieces Density: 5.85 Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447


For vacuum deposition

Formula: BaO₃Ti
Melting Pt: 1620 °C
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR