Mercury(II) telluride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

88244-06
CAAA88244-06EA 312.84 CAD
CAAA88244-06 CAAA88244-14
Mercury(II) telluride ≥99% (metals basis)
Mercury (II) telluride

MDL: MFCD00016133


Purity: 99% CAS No.: 12068-90-5 Molecular Formula: HgTe Formula Weight: 328.19 Physical Form: -100 Mesh Powder Density: 8.63 Appearance: Gray to black Odor: Odorless Melting Point: 673° UN No.: 2025 MDL No.: MFCD00016133

Formula: HgTe
Melting Pt: 673 °C
Density: 8,63
Storage Temperature: Ambient
MDL Number: MFCD00016133
CAS Number: 12068-90-5
EINECS: 235-108-9
UN: 2025
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR