Molecular sieve A5 (0.5 nm, 5 Å), pellets, Ø 1-2mm (0.04-0.08in)

Supplier: Thermo Scientific Chemicals

87955-30
CAAA87955-30EA 49.93 CAD
CAAA87955-30
Molecular sieve A5 (0.5 nm, 5 Å), pellets, Ø 1-2mm (0.04-0.08in)
Molecular sieve A5 (0.5 nm, 5 Å)

Pellets


CAS No.: 69912-79-4 Physical Form: Pellets Sensitivity: Hygroscopic Appearance: Pale brown MDL No.: MFCD00147627

MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR