Butyltin trichloride 96

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Butyltin trichloride, Trichlorobutylstannane

71125-14 71125-22
CAAA71125-14EA 52.14 CAD
CAAA71125-14 CAAA71125-22
Butyltin trichloride 96
Butyltin trichloride

MDL: MFCD00000515


Purity: 96% CAS No.: 1118-46-3 Molecular Formula: CH3(CH2)3SnCl3 Formula Weight: 282.17 Flash Point: 81°(178°F) Physical Form: Liquid Density: 1.693 Refractive Index: 1.5244 Sensitivity: Moisture Sensitive Odor: Acrid Melting Point: -63° Boiling Point: 98°/10mm UN No.: 3265 MDL No.: MFCD00000515

Formula: C₄H₉Cl₃Sn
MW: 282.18
Boiling Pt: 227
Melting Pt: –63 °C
Density: 1.693
Flash Pt: 81 °C (178 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000515
CAS Number: 1118-46-3
EINECS: 214-263-6
UN: 3265
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR