Disodium sulfide, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium sulfide, Sodium sulphide

65122-06 65122-14 65122-22
CAAA65122-06EA 111.55 CAD
CAAA65122-06 CAAA65122-14 CAAA65122-22
Disodium sulfide, anhydrous
Disodium sulphide

In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagent


CAS No.: 1313-82-2 Molecular Formula: Na2S Formula Weight: 78.04 Physical Form: Granular Density: 1.856 Sensitivity: Light Sensitive and Hygroscopic Appearance: Pale yellow Odor: Like rotten eggs Melting Point: 950° Solubility: Soluble in water. Decomposes in acid. Slightly soluble in alcohol. Insoluble in ether Merck Reference: 14,8681 UN No.: 1385 MDL No.: MFCD00003498


In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagent

Formula: Na₂S
MW: 78.05 g/mol
Boiling Pt: 174
Melting Pt: 950 °C
Density: 1.856
Storage Temperature: Ambient
MDL Number: MFCD00003498
CAS Number: 1313-82-2
EINECS: 215-211-5
UN: 1385
ADR: 4.2 II
REACH: 01-2119513694-38
Merck Index: 14,8681

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR