Potassium molybdate(VI) ≥95%

Supplier: Thermo Scientific Chemicals
Warning

48112-22 48112-36
CAAA48112-22EA 122.45 CAD
CAAA48112-22 CAAA48112-36
Potassium molybdate(VI) ≥95%
Potassium molybdate(VI)

MDL: MFCD00011370 Soluble in water. Insoluble in alcohol


Purity: 95% CAS No.: 13446-49-6 Molecular Formula: K2MoO4 Formula Weight: 238.14 Physical Form: Powder Density: 2.3 Melting Point: 919° Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,7647 MDL No.: MFCD00011370

Formula: K₂MoO₄
Melting Pt: 919 °C
Density: 2.3
Storage Temperature: Ambient
MDL Number: MFCD00011370
CAS Number: 13446-49-6
EINECS: 236-599-2
Merck Index: 14,7647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR