Inconel® 600 alloy, gauze woven from wire 60 mesh Ø 0.19 mm (0.0075 in)
Supplier: Thermo Scientific Chemicals
Danger
46584-HB
CAAA46584-FLEA 47.27 CAD
CAAA46584-FL CAAA46584-GT CAAA46584-HB
Inconel® 600 alloy, gauze woven from wire 60 mesh Ø 0.19 mm (0.0075 in)
Inconel® 600 alloy
Wire Cloth
Molecular Formula: Ni:Cr:Fe; 77:15.5:7.5 wt% Physical Form: Wire Cloth MDL No.: MFCD01741927