Trisodium citrate, anhydrous 99%

Supplier: Thermo Scientific Chemicals

45556-A7
CAAA45556-A7EA 687.3 CAD
CAAA45556-A7
Trisodium citrate, anhydrous 99%
tri-Sodium citrate

Purity: 99% CAS No.: 68-04-2 Molecular Formula: C6H5Na3O7 Formula Weight: 258.07 Physical Form: Solid Sensitivity: Hygroscopic Appearance: White Melting Point: 300° Merck Reference: 14,8602 MDL No.: MFCD00012462

Formula: C₆H₅Na₃O₇
Melting Pt: 300 °C
Density: 1.815 g/cm³ (20 °C)
Flash Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012462
CAS Number: 68-04-2
EINECS: 200-675-3
Merck Index: 14,8602

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR