Magnesium iodide, ultra dry ≥99.996% (metals basis)

Supplier: Thermo Scientific Chemicals

44310-14 44310-22
CAAA44310-14EA 750.5 CAD
CAAA44310-14 CAAA44310-22
Magnesium iodide, ultra dry ≥99.996% (metals basis)
Magnesium iodide

MDL: MFCD00049485


Purity: 99.996% CAS No.: 10377-58-9 Molecular Formula: I2Mg Formula Weight: 278.12 Physical Form: Powder, Ampouled under argon Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odor: Odorless Melting Point: 637° dec. Merck Reference: 14,5671 MDL No.: MFCD00049485

Formula: MgI₂
Melting Pt: 637 °C (dec.)
Density: 3.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049485
CAS Number: 10377-58-9
EINECS: 233-825-1
Merck Index: 14,5671

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR