Silicon 99,9 Metals basis, powder APS 1-5 µm

Supplier: Thermo Scientific Chemicals
Warning

44185-09 44185-18 44185-30
CAAA44185-09EA 144.41 CAD
CAAA44185-09 CAAA44185-18 CAAA44185-30
Silicon 99,9 Metals basis, powder APS 1-5 µm
Silicon

Purity: 99.9% CAS No.: 7440-21-3 Physical Form: Powder Appearance: White Odor: Odorless UN No.: 1346 MDL No.: MFCD00085311

Formula: Si
MW: 28.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1 III
Merck Index: 12,08635

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR