Gold ≥99.999% (metals basis), foil, Premion®, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

44163-FF 44163-FI
CAAA44163-FFEA 1358.8 CAD
CAAA44163-FF CAAA44163-FI
Gold ≥99.999% (metals basis), foil, Premion®, Thickness: 0.25 mm (0.01 in)
Gold

MDL: MFCD00003436


Purity: 99.999% Physical Form: ≈3g/25x25mm Odor: Odorless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR