Lithium iodide, ultra dry ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

44159-09 44159-18 44159-30
CAAA44159-09EA 99.07 CAD
CAAA44159-09 CAAA44159-18 CAAA44159-30
Lithium iodide, ultra dry ≥99% (metals basis)
Lithium iodide

MDL: MFCD00011092


Purity: 99% CAS No.: 10377-51-2 Molecular Formula: ILi Formula Weight: 133.84 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.076 Refractive Index: 1.955 Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odor: Odorless Melting Point: 449° Boiling Point: 1170-1190° Merck Reference: 14,5535 MDL No.: MFCD00011092

Formula: ILi
Boiling Pt: 1170…1190 °C
Melting Pt: 449 °C
Density: 4.076
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Merck Index: 14,5535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR