MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 98%

Supplier: Thermo Scientific Chemicals
Warning

44135-18 44135-30 44135-A1
CAAA44135-09EA 42.94 CAD
CAAA44135-09 CAAA44135-18 CAAA44135-30 CAAA44135-A1
MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 98%
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)

Biological buffers, pH range 6.2-7.6


Purity: 98% CAS No.: 68399-77-9 Molecular Formula: C7H15NO5S Formula Weight: 225.27 Physical Form: Powder Melting Point: 276-279° MDL No.: MFCD00009629


Biological buffers, pH range 6.2-7.6

Formula: C₇H₁₅NO₅S
Melting Pt: 276…279 °C
Storage Temperature: Ambient
MDL Number: MFCD00009629
CAS Number: 68399-77-9
EINECS: 269-989-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR