3-Phenyl-2-propyn-1-ol 98+%

Supplier: Thermo Scientific Chemicals

43778-04 43778-09 43778-18
CAAA43778-04EA 62.09 CAD
CAAA43778-04 CAAA43778-09 CAAA43778-18
3-Phenyl-2-propyn-1-ol 98+%
3-Phenyl-2-propyn-1-ol

MDL: MFCD00040914 Beilstein Registry No.: 2040911


Purity: 98+% CAS No.: 1504-58-1 Molecular Formula: C6H5C≡CCH2OH Formula Weight: 132.16 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.078 Refractive Index: 1.5867 Boiling Point: 129-130°/10mm Beilstein Reference: 2040911 MDL No.: MFCD00040914

Formula: C₉H₈O
Boiling Pt: 129…130 °C (10 mmHg)
Density: 1,078
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040914
CAS Number: 1504-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR