Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)

Supplier: Thermo Scientific Chemicals

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

43728-18 43728-30 43728-A1
CAAA43728-18EA 62.85 CAD
CAAA43728-18 CAAA43728-30 CAAA43728-A1
Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)
Polyvinylpyrrolidone

MDL: MFCD00149016


CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Physical Form: Powder Sensitivity: Hygroscopic Odor: Odorless Merck Reference: 14,7697 MDL No.: MFCD00149016

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD00149016
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR