2,3-Dimethylpentane 97%

Supplier: Thermo Scientific Chemicals
Danger

43707-06 43707-14
CAAA43707-06EA 97.96 CAD
CAAA43707-06 CAAA43707-14
2,3-Dimethylpentane 97%
2,3-Dimethylpentane

Purity: 97% CAS No.: 565-59-3 Molecular Formula: C7H16 Formula Weight: 100.21 Flash Point: -6°(21°F) Physical Form: Liquid Density: 0.690 Refractive Index: 1.3920 Boiling Point: 89-90° UN No.: 1206 MDL No.: MFCD00009337

Formula: C₇H₁₆
Boiling Pt: 89…90 °C
Melting Pt: –135 °C
Density: 0.690
Flash Pt: –6 °C (21 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009337
CAS Number: 565-59-3
EINECS: 209-280-0
UN: 1206
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR