Calcium iodide, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

43674-14 43674-22 43674-36
CAAA43674-14EA 251.22 CAD
CAAA43674-14 CAAA43674-22 CAAA43674-36
Calcium iodide, ultra dry ≥99.99% (metals basis)
Calcium iodide

MDL: MFCD00010910


Purity: 99.99% CAS No.: 10102-68-8 Molecular Formula: CaI2 Formula Weight: 293.89 Physical Form: Powder, Ampouled under argon Density: 3.956 Appearance: White to pale yellow Odor: Odorless Melting Point: 784° Boiling Point: 1100° Merck Reference: 14,1677 MDL No.: MFCD00010910

Formula: CaI₂
Boiling Pt: 1100 °C
Melting Pt: 784 °C
Density: 3.956
Storage Temperature: Ambient
MDL Number: MFCD00010910
CAS Number: 10102-68-8
EINECS: 233-276-8
Merck Index: 14,1677

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR