Glycine ≥98.5% ACS

Supplier: Thermo Scientific Chemicals

43497-22 43497-36 43497-A5
CAAA43497-22EA 73.94 CAD
CAAA43497-22 CAAA43497-36 CAAA43497-A5
Glycine ≥98.5% ACS
Glycine

MDL: MFCD00008131 Beilstein Registry No.: 635782 Fieser: 1,412


Purity: 98.5+% CAS No.: 56-40-6 Molecular Formula: NH2CH2CO2H Formula Weight: 75.07 Physical Form: Crystalline Density: 1.595 Appearance: White to off-white Odor: Odorless pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: ∼ 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Residue after ignition 0.1
Heavy metals (as Pb) 0.2% (as Pb)
Cl 0.005 AU
SO3 0.005%
NH3 0.005%
Substances darkened by sulfuric acid Pass test
Hydrolyzable substances Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR