Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%

Supplier: Thermo Scientific Chemicals
Warning

43384-03 43384-06
CAAA43384-03EA 138.57 CAD
CAAA43384-03 CAAA43384-06
Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%
Methyl-4-aminothiophene-3-carboxylate hydrochloride

Purity: 97+% CAS No.: 39978-14-8 Molecular Formula: C6H7NO2S.HCl Formula Weight: 193.65 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Brown Melting Point: 199-200° dec. MDL No.: MFCD00068149

Formula: C₆H₈ClNO₂S
Melting Pt: 199…200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00068149
CAS Number: 39978-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR