Zirconyl nitrate hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

43224-14 43224-22 43224-36
CAAA43224-14EA 127.82 CAD
CAAA43224-14 CAAA43224-22 CAAA43224-36
Zirconyl nitrate hydrate ≥99.9% (metals basis)
Zirconyl nitrate hydrate

Purity: 99.9% CAS No.: 14985-18-3 Molecular Formula: ZrO(NO3)2.xH2O Formula Weight: 231.23(anhy) Physical Form: Crystalline Odor: Odorless UN No.: 1477 MDL No.: MFCD00149896

Formula: N₂O₇Zr·H₂O
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR