Lanthanum(III) sulfate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

43106-22 43106-36
CAAA43106-22EA 161.16 CAD
CAAA43106-22 CAAA43106-36
Lanthanum(III) sulfate hydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum (III) sulphate hydrate

MDL: MFCD00011070


Purity: 99.9% CAS No.: 57804-25-8 Molecular Formula: La2(SO4)3.xH2O Formula Weight: 566.00(anhy) Physical Form: Crystalline Density: 2.821 Melting Point: dec. white heat Merck Reference: 14,5363 MDL No.: MFCD00011070

Formula: La₂O₁₂S₃·H₂O
Melting Pt: Dec. (white heat)
Density: 2.821
Storage Temperature: Ambient
MDL Number: MFCD00011070
CAS Number: 57804-25-8
EINECS: 233-239-6
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR