Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

42982-BU 42982-BY 42982-CC
CAAA42982-BUEA 71.42 CAD
CAAA42982-BU CAAA42982-BY CAAA42982-CC
Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.5 mm (0.02 in)
Hafnium

MDL: MFCD00011032


Purity: 99.95% CAS No.: 7440-58-6 Physical Form: ≈2.6g/m Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR