Cadmium(II) chloride, ultra dry ≥99.996% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

42852-04 42852-09 42852-18 42852-30
CAAA42852-04EA 55.93 CAD
CAAA42852-04 CAAA42852-09 CAAA42852-18 CAAA42852-30
Cadmium(II) chloride, ultra dry ≥99.996% (metals basis)
Cadmium (II) chloride

MDL: MFCD00010916


Purity: 99.996% CAS No.: 10108-64-2 Molecular Formula: CdCl2 Formula Weight: 183.32 Physical Form: Powder, Ampouled under argon Density: 4.047 Melting Point: 568° Boiling Point: 960° Merck Reference: 14,1617 UN No.: 2570 MDL No.: MFCD00010916

Formula: CdCl₂
Boiling Pt: 960 °C
Melting Pt: 568 °C
Density: 4.047
Storage Temperature: Ambient
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 14,1617

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR