Nickel chromium alloy (80:20 w%), wire, Ø 1.6 mm (0.064 in)

Supplier: Thermo Scientific Chemicals
Warning

42838-G6
CAAA42838-G6EA 136.67 CAD
CAAA42838-G6 CAAA42838-H2
Nickel chromium alloy (80:20 w%), wire, Ø 1.6 mm (0.064 in)
Nickel Chromium alloy (80:20 w%)

MDL: MFCD00673110


CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: ≈17g/m Odor: Odorless MDL No.: MFCD00673110

Storage Temperature: Ambient
MDL Number: MFCD00673110
CAS Number: 11106-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR