Antimony(III) iodide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

42831-04 42831-09 42831-18
CAAA42831-04EA 115.66 CAD
CAAA42831-04 CAAA42831-09 CAAA42831-18
Antimony(III) iodide, ultra dry ≥99.999% (metals basis)
Antimony (III) iodide

MDL: MFCD00016322


Purity: 99.999% CAS No.: 7790-44-5 Molecular Formula: SbI3 Formula Weight: 502.46 Physical Form: Powder, Ampouled under argon Sensitivity: Moisture Sensitive Boiling Point: 420° Merck Reference: 14,710 UN No.: 3260 MDL No.: MFCD00016322

Formula: I₃Sb
Boiling Pt: 420 °C
Melting Pt: 168 °C
Density: 4.676 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016322
CAS Number: 7790-44-5
EINECS: 232-205-8
UN: 3077
ADR: 9,III
Merck Index: 14,710

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR