Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 4.5 micron

Supplier: Thermo Scientific Chemicals

42715-AA
CAAA42715-AAEA 1012.78 CAD
CAAA42715-AA
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 4.5 micron
Poly(styrene)

CAS No.: 9003-53-6 Molecular Formula: [-CH2CH(C6H5)-]n Storage Temperature: 2°C to 8°C Physical Form: Dispersion Appearance: Opalescent Odor: Odorless MDL No.: MFCD00084450

Formula: C₈H₈
Storage Temperature: Refrigerator
MDL Number: MFCD00084450
CAS Number: 9003-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR