Strontium titanate substrate 5x5x0.5 mm, polished one side 100 orientation

Supplier: Thermo Scientific Chemicals

42620-KT
CAAA42620-KTEA 206.98 CAD
CAAA42620-KT CAAA42620-KU
Strontium titanate substrate 5x5x0.5 mm, polished one side 100 orientation
Strontium titanate

MDL: MFCD00049554


CAS No.: 12060-59-2 Molecular Formula: SrTiO3 Odor: Odorless MDL No.: MFCD00049554

Formula: O₃SrTi
MW: 183.5
Melting Pt: 2080
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR