Sulfuric acid 15% (v/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

42554-K2
CAAA42554-K2EA 90.53 CAD
CAAA42554-K2
Sulfuric acid 15% (v/v) in aqueous solution
Sulfuric acid

Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid UN No.: 2796 MDL No.: MFCD00064589

Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR