Inhibitor removal resin

Supplier: Thermo Scientific Chemicals

42489-14 42489-22 42489-36
CAAAA42489-14EA 52.46 CAD
CAAAA42489-14 CAAA42489-22 CAAA42489-36
Inhibitor removal resin
Inhibitor removal resin

Applications: For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol


CAS No.: 9003-70-7 Physical Form: Crystalline Odor: Odorless MDL No.: MFCD00081548


Applications: For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol

Storage Temperature: Ambient
MDL Number: MFCD00081548
CAS Number: 9003-70-7

Order Now

Specification Test Results

Form Crystalline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR