2,2,2-Trifluoroethanol-[D₃] (99% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

42363-03 42363-46
CAAA42363-03EA 244.9 CAD
CAAA42363-03 CAAA42363-46
2,2,2-Trifluoroethanol-[D₃] (99% D) (Isotopic) for NMR spectroscopy
2,2,2-Trifluoroethanol-[D3]

NMR Solvent


Purity: 99% CAS No.: 77253-67-9 Molecular Formula: CF3CD2OD Formula Weight: 103.06 Physical Form: Liquid in prescored ampoules Density: 1.45 Melting Point: -44° Boiling Point: 77-80° UN No.: 1992 MDL No.: MFCD00037682


NMR Solvent

Formula: F₃CCD₂OD
Boiling Pt: 77…80 °C
Melting Pt: –44 °C
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00037682
CAS Number: 77253-67-9
EINECS: 278-649-6
UN: 1992
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR