Nitromethane-[D₃] (99% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

42338-06
CAAA42338-06EA 147.1 CAD
CAAA42338-06
Nitromethane-[D₃] (99% D) (Isotopic) for NMR spectroscopy
Nitromethane-[D3]

MDL: MFCD00044214


Purity: 99% CAS No.: 13031-32-8 Molecular Formula: CD3NO2 Formula Weight: 64.06 Physical Form: Liquid Density: 1.183 Refractive Index: 1.3795 Appearance: Colorless Odor: Characteristic Boiling Point: 100° UN No.: 1261 MDL No.: MFCD00044214

Formula: D₃CNO₂
Boiling Pt: 100 °C
Melting Pt: –26 °C
Density: 1.183
MDL Number: MFCD00044214
CAS Number: 13031-32-8
EINECS: 235-892-2
UN: 1261
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR